1,4-Dihydroxy-2-naphthoic acid, 98%, Thermo Scientific™
1,4-Dihydroxy-2-naphthoic acid, 98%, Thermo Scientific™
1,4-Dihydroxy-2-naphthoic acid, 98%, Thermo Scientific™
1,4-Dihydroxy-2-naphthoic acid, 98%, Thermo Scientific™
1,4-Dihydroxy-2-naphthoic acid, 98%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFH60210.1425 g
Catalog number ALFH60210.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1,4-Dihydroxy-2-naphthoic acid
CAS31519-22-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1,4-Dihydroxy-2-naphthoic acid from Propionibacterium freudenreichii is known to promote the proliferation of Bifidobacterium. It has potential therapeutic application for psoriasis treatment.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
General References:
  1. Yoshikiyo Okada; Yoshikazu Tsuzuki; Kazuyuki Narimatsu; Hirokazu Sato; Toshihide Ueda; Hideaki Hozumi; Shingo Sato; Ryota Hokari; Chie Kurihara; Shunsuke Komoto; Chikako Watanabe; Kengo Tomita; Atsushi Kawaguchi; Shigeaki Nagao; Soichiro Miura. 1,4-Dihydroxy-2-naphthoic acid from Propionibacterium freudenreichii reduces inflammation in interleukin-10-deficient mice with colitis by suppressing macrophage-derived proinflammatory cytokines. Journal of Leukocyte Biology. 2013, 94,(3), 473-480.
  2. Chong-Fai Mok; Chuan-Ming Xie; Kathy Wai-Yan Sham; Zhi-Xiu Lin; Christopher Hon-Ki Cheng. 1,4-dihydroxy-2-naphthoic acid induces apoptosis in human keratinocyte: Potential application for psoriasis treatment. Evidence-based Complementary and Alternative Medicine. 2013, 2013,792840.