Asiatic acid, 97%
Asiatic acid, 97%
Asiatic acid, 97%
Thermo Scientific Chemicals
Asiatic acid, 97%
CAS: 464-92-6 | C30H48O5 | 488.709 g/mol
Have Questions?
Catalog NumberQuantity
ALFH60149.ME500 mg
Catalog number ALFH60149.ME
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Request bulk or custom format
Chemical Identifiers
CAS99-77-4
IUPAC Nameethyl 4-nitrobenzoate
Molecular FormulaC9H9NO4
InChI KeyPHWSCBWNPZDYRI-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point56°C to 59°C
GC>=98.5 %
Asiatic acid is commonly used in wound healing. Asiatic acid has antioxidant, anti-inflammatory and neuroprotective properties. It is a starting material for asiatic acid derivative synthesis for use as anticancer agents, glycogen phosphorylase inhibitors and and hepatoprotectants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Asiatic acid is commonly used in wound healing. Asiatic acid has antioxidant, anti-inflammatory and neuroprotective properties. It is a starting material for asiatic acid derivative synthesis for use as anticancer agents, glycogen p
RUO – Research Use Only
Kyung-Jin Yun.; Ji-Yeon Kim.; Jong-Bin Kim.; Kyung-Won Lee.; Seo-Young Jeong.; Hee-Juhn Park.; Hyun-Ju Jung.; Young-Wuk Cho.; Kijoo Yun.; Kyung-Tae Lee. Inhibition of LPS-induced NO and PGE2 production by asiatic acid via NF-κB inactivation in RAW 264.7 macrophages: Possible involvement of the IKK and MAPK pathways.International Immunopharmacology. 2008, 8 (3),431-441 .