Thiazole-5-carboxaldehyde, 95%, Thermo Scientific™
Thiazole-5-carboxaldehyde, 95%, Thermo Scientific™
Thiazole-5-carboxaldehyde, 95%, Thermo Scientific™
Thiazole-5-carboxaldehyde, 95%, Thermo Scientific™
Thiazole-5-carboxaldehyde, 95%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFH60138.031 g
Catalog number ALFH60138.03
Price (MYR)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialThiazole-5-carboxaldehyde
CAS1003-32-3
Health Hazard 1H302-H317-H319
Health Hazard 2GHS H Statement
H301-H319-H317
Toxic if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.