kappa-Carrageenan, Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
Thermo Scientific Chemicals
kappa-Carrageenan, Thermo Scientific Chemicals
CAS: 11114-20-8 | C24H36O25S2-2 | 788.647 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFH60117.14
Price (MYR)
1,293.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,293.00
Add to cart
Specifications
Chemical Name or Materialkappa-Carrageenan
CAS11114-20-8
Health Hazard 1H332
Health Hazard 3P261-P271-P304+P340-P312