1-n-Butyl-3-methylimidazolium di-n-butyl phosphate, 96%, Thermo Scientific Chemicals
1-n-Butyl-3-methylimidazolium di-n-butyl phosphate, 96%, Thermo Scientific Chemicals
1-n-Butyl-3-methylimidazolium di-n-butyl phosphate, 96%, Thermo Scientific Chemicals
1-n-Butyl-3-methylimidazolium di-n-butyl phosphate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-n-Butyl-3-methylimidazolium di-n-butyl phosphate, 96%, Thermo Scientific Chemicals
CAS: 663199-28-8 | C16H33N2O4P | 348.424 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
Catalog number ALFH59893.18
Price (MYR)
2,685.00
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
2,685.00
Add to cart
Specifications
Chemical Name or Material1-n-Butyl-3-methylimidazolium di-n-butyl phosphate
CAS663199-28-8
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
1-Butyl-3-methylimidazolium dibutyl phosphate is employed as an imidazolium compound salt.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Butyl-3-methylimidazolium dibutyl phosphate is employed as an imidazolium compound salt.
Solubility
Insoluble in water.
Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Vincenzo Calò.;Angelo Nacci .; Antonio Monopoli.; Antonio Fornaro.; Luigia Sabbatini.; Nicola Cioffi.; Nicoletta Ditaranto. Heck Reaction Catalyzed by Nanosized Palladium on Chitosan in Ionic Liquids.Organometallics. 2004, 23 (22),5154-5158.