1-Allyl-3-methylimidazolium bromide, 97%, Thermo Scientific Chemicals
1-Allyl-3-methylimidazolium bromide, 97%, Thermo Scientific Chemicals
1-Allyl-3-methylimidazolium bromide, 97%, Thermo Scientific Chemicals
1-Allyl-3-methylimidazolium bromide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Allyl-3-methylimidazolium bromide, 97%, Thermo Scientific Chemicals
CAS: 31410-07-8 | C7H11BrN2 | 203.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFH59513.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS35141-30-1
IUPAC Name3,3-dimethoxy-2-oxa-7,10-diaza-3-siladodecan-12-amine
Molecular FormulaC10H27N3O3Si
InChI KeyNHBRUUFBSBSTHM-UHFFFAOYSA-N
SMILESCO[Si](CCCNCCNCCN)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4570 to 1.4650 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
AppearanceMay discolor to pink to red to brown on storage