N-Fmoc-S-acetamidomethyl-L-cysteine, 95%, Thermo Scientific Chemicals
N-Fmoc-S-acetamidomethyl-L-cysteine, 95%, Thermo Scientific Chemicals
N-Fmoc-S-acetamidomethyl-L-cysteine, 95%, Thermo Scientific Chemicals
N-Fmoc-S-acetamidomethyl-L-cysteine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Fmoc-S-acetamidomethyl-L-cysteine, 95%, Thermo Scientific Chemicals
CAS: 86060-81-3 | C21H22N2O5S | 414.48 g/mol
Have Questions?
Catalog NumberQuantity
ALFH59306.065 g
Catalog number ALFH59306.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1309-64-4
IUPAC Namediantimony(3+) trioxidandiide
Molecular FormulaO3Sb2
InChI KeyGHPGOEFPKIHBNM-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Sb+3].[Sb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.