1-Ethyl-3-methylimidazolium acetate, 97%, Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium acetate, 97%, Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium acetate, 97%, Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium acetate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium acetate, 97%, Thermo Scientific Chemicals
CAS: 143314-17-4 | C8H14N2O2 | 170.212 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number ALFH59277.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Ethyl-3-methylimidazolium acetate
CAS143314-17-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is used as a solvent and as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a solvent and as pharmaceutical intermediate.
Solubility
Soluble in water, acetonitrile, acetone, dichloromethane.
Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Ning Sun.; Mustafizur Rahman.; Ying Qin.; Mirela L. Maxim.; Héctor Rodríguez.; Robin D. Rogers. Complete dissolution and partial delignification of wood in the ionic liquid 1-ethyl-3-methylimidazolium acetate.Green Chem. 2009, 11 (5),646-655.
  2. Jing-Fang Huang.; Po-Yu Chen.; I-Wen Sun.; S.P. Wang. NMR evidence of hydrogen bonding in 1-ethyl-3-methylimidazolium-tetrafluoroborate room temperature ionic liquid. Inorg. Chim. Acta. 2001, 320 (1-2), 7-11.