Nepsilon-Boc-Nalpha-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals
Nepsilon-Boc-Nalpha-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals
Nepsilon-Boc-Nalpha-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals
Nepsilon-Boc-Nalpha-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nepsilon-Boc-Nalpha-Fmoc-L-lysine, 98%, Thermo Scientific Chemicals
CAS: 71989-26-9 | C26H32N2O6 | 468.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFH59182.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNepsilon-Boc-Nalpha-Fmoc-L-lysine
CAS71989-26-9
Melting Point130°C to 135°C
Recommended StorageKeep cold
EINECS Number276-256-4
View more
It is used to prepare dimeric RGD peptide-paclitaxel conjugate as model for integrin-targeted drug delivery. It is also used to synthesize DOTA-conjugated multivalent cyclic-RGD peptide dendrimers for tumor targeting and imaging use.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used to prepare dimeric RGD peptide-paclitaxel conjugate as model for integrin-targeted drug delivery. It is also used to synthesize DOTA-conjugated multivalent cyclic-RGD peptide dendrimers for tumor targeting and imaging use.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Xiaoyuan Chen; Carmen Plasencia; Yingping Hou; and Nouri Neamati. Synthesis and Biological Evaluation of Dimeric RGD Peptide-Paclitaxel Conjugate as a Model for Integrin-Targeted Drug Delivery. J. Med. Chem,2005,48 (4), 1098-1106.
  2. Dijkgraaf, I., et al. Synthesis of DOTA-conjugated multivalent cyclic-RGD peptide dendrimers via 1,3-dipolar cycloaddition and their biological evaluation: implications for tumor targeting and tumor imaging purposes. Org. Biomol. Chem,2007, 5(6), 935-944.