N-Fmoc-L-glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals
N-Fmoc-L-glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals
N-Fmoc-L-glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals
N-Fmoc-L-glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Fmoc-L-glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals
CAS: 71989-18-9 | C24H27NO6 | 425.481 g/mol
Have Questions?
Catalog NumberQuantity
ALFH59169.1425 g
Catalog number ALFH59169.14
Price (MYR)
1,396.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,396.00
Add to cart
Specifications
Chemical Name or MaterialN-Fmoc-L-glutamic acid 5-tert-butyl ester
CAS71989-18-9
Melting Point83°C to 90°C
Recommended StorageKeep cold
EINECS Number276-253-8