4-(Trifluoromethyl)-1-indanone, 97%, Thermo Scientific Chemicals
4-(Trifluoromethyl)-1-indanone, 97%, Thermo Scientific Chemicals
4-(Trifluoromethyl)-1-indanone, 97%, Thermo Scientific Chemicals
4-(Trifluoromethyl)-1-indanone, 97%, Thermo Scientific Chemicals
4-(Trifluoromethyl)-1-indanone, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFH59165.06
Price (MYR)
934.65
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
934.65
Add to cart
Specifications
Chemical Name or Material4-(Trifluoromethyl)-1-indanone
CAS68755-42-0
Health Hazard 1H302-H317
Health Hazard 2GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c