N-Fmoc-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals
N-Fmoc-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals
N-Fmoc-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals
N-Fmoc-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Fmoc-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals
CAS: 71989-14-5 | C23H25NO6 | 411.454 g/mol
Have Questions?
Catalog NumberQuantity
ALFH59007.1425 g
Catalog number ALFH59007.14
Price (MYR)
1,347.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,347.00
Add to cart
Specifications
Chemical Name or MaterialN-Fmoc-L-aspartic acid 4-tert-butyl ester
CAS71989-14-5
Melting Point148°C to 150°C (decomposition)
Recommended StorageKeep cold
EINECS Number276-251-7
View more
L-Aspartic acid is a nonessential amino acid that is used to biosynthesize other amino acids within the human body.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Aspartic acid is a nonessential amino acid that is used to biosynthesize other amino acids within the human body.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Anderson, H., et al. Effect on Nitrogen Balance of Adult Man of Varying Source of Nitrogen and Level of Calorie Intake. J. Nutr.,1969, 99(1), 82-90.
  2. C.R. Honey; Z. Miljkovic; J.F. Macdonald. Ketamine and phencyclidine cause a voltage-dependent block of responses to l-aspartic acid. Neuroscience Letters,1985, 61(1-2), 135-139.