1,6-Bismaleimidohexane, 97%, Thermo Scientific Chemicals
1,6-Bismaleimidohexane, 97%, Thermo Scientific Chemicals
1,6-Bismaleimidohexane, 97%, Thermo Scientific Chemicals
1,6-Bismaleimidohexane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,6-Bismaleimidohexane, 97%, Thermo Scientific Chemicals
CAS: 4856-87-5 | C14H16N2O4 | 276.292 g/mol
Have Questions?
Catalog NumberQuantity
ALFH58874.MC100 mg
Catalog number ALFH58874.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Specifications
Chemical Name or Material1,6-Bismaleimidohexane
CAS4856-87-5
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
A sulfhydryl reactive homobifunctional cross linking reagent. Under mild conditions, will permit irreversible cross-linking of sulfhydral containing compounds at a pH range of 6.5 to 7.5.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A sulfhydryl reactive homobifunctional cross linking reagent. Under mild conditions, will permit irreversible cross-linking of sulfhydral containing compounds at a pH range of 6.5 to 7.5.
Solubility
Soluble in chloroform.
Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only
Naozumi Teramoto.; Yohei Arai.; Mitsuhiro Shibata. Thermo-reversible Diels-Alder polymerization of difurfurylidene trehalose and bismaleimides. Carbohydr. Polym. 2006, 64 (1),78-84.