9,9-Di-n-hexyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
9,9-Di-n-hexyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
9,9-Di-n-hexyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
9,9-Di-n-hexyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
9,9-Di-n-hexyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
CAS: 189367-54-2 | C25H32Br2 | 492.339 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFH56952.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS4848-43-5
IUPAC Name(2-aminoethyl)diphenylphosphane
Molecular FormulaC14H16NP
InChI KeyRXEPBCWNHKZECN-UHFFFAOYSA-N
SMILESNCCP(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥94.0% (UK sourced material)
Assay from Supplier's CofA≥94.0% (GC, US sourced material)
Assay from Supplier's CofA≥94.0% (P31-NMR, US sourced material)
View more
It finds its application as an intermediate for polymeric light-emitting diodes and as an OLED material because of their pure blue emission and efficient electroluminescence coupled with a high charge-carrier mobility and good process ability.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application as an intermediate for polymeric light-emitting diodes and as an OLED material because of their pure blue emission and efficient electroluminescence coupled with a high charge-carrier mobility and good process ability.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. V. N. Bliznyuk and S. A. Carter. Electrical and Photo induced Degradation of Polyfluorene Based Films and Light-Emitting Devices.Macromolecules.,1999,32(2), 361-369.
  2. C. M. Martin, et al. Hydrostatic pressure dependence of the luminescence and Raman frequencies in polyfluorene.Phys. Rev. B.,2003,68(11), 115203.