Phenazine methosulfate, 98+%, Thermo Scientific Chemicals
Phenazine methosulfate, 98+%, Thermo Scientific Chemicals
Phenazine methosulfate, 98+%, Thermo Scientific Chemicals
Phenazine methosulfate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenazine methosulfate, 98+%, Thermo Scientific Chemicals
CAS: 299-11-6 | C14H14N2O4S | 306.336 g/mol
Have Questions?
Catalog NumberQuantity
ALFH56718.065 g
Catalog number ALFH56718.06
Price (MYR)
985.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
985.00
Add to cart
Specifications
Chemical Name or MaterialPhenazine methosulfate
CAS299-11-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is used with ascorbic acid to determine nitric oxide reductase activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used with ascorbic acid to determine nitric oxide reductase activity.
Solubility
Soluble in water. (100g/L at20°C)
Notes
Light sensitive. Incompatible with oxidizing agents. Store at -20°C.
RUO – Research Use Only
General References:
  1. Morimitsu Nishikimi.; N. Appaji Rao.; Kunio Yagi. The occurrence of superoxide anion in the reaction of reduced phenazine methosulfate and molecular oxygen. Biochem. Biophys. Res. Commun. 1972, 46 (31),849-854.
  2. Wilhelmus N. Konings.; Eugene M. Barnes Jr.; H. R. Kaback. Mechanisms of Active Transport in Isolated Membrane Vesicles III. The Coupling Of Reduced Phenazine Methosulfate To The Concentrative Uptake Of Β-Galactosides And Amino Acids.The Journal of Biological Chemistry. 1971, 2465857-5861.