9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
CAS: 286438-45-7 | C37H56Br2 | 660.663 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFH56320.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS67-66-3
IUPAC Nametrichloromethane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-UHFFFAOYSA-N
SMILESClC(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at245nm: 1.00AU, max
FormColorless liquid
Color (APHA)10, max
CommentSubmicron filtered
Impurity contentAcetone and aldehyde ( as acetone): Passes Test
View more
It is an intermediate for polymeric light-emitting diodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an intermediate for polymeric light-emitting diodes.
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
RUO – Research Use Only