9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
9,9-Di-n-dodecyl-2,7-dibromofluorene, 98%, Thermo Scientific Chemicals
CAS: 286438-45-7 | C37H56Br2 | 660.663 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFH56320.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material9,9-Di-n-dodecyl-2,7-dibromofluorene
CAS286438-45-7
Melting Point50°C to 55°C
Recommended StorageAmbient temperatures
TSCANo
View more
It is an intermediate for polymeric light-emitting diodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an intermediate for polymeric light-emitting diodes.
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
RUO – Research Use Only