3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 98+%, Thermo Scientific Chemicals
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 98+%, Thermo Scientific Chemicals
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 98+%, Thermo Scientific Chemicals
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 98+%, Thermo Scientific Chemicals
CAS: 38894-11-0 | C8H12ClN3OS | 233.714 g/mol
Have Questions?
Change viewbuttonViewtableView
Catalog number ALFH56061.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight233.72 (215.70 Anhydrous)
Unit Size
It is used as a reagent for the spectrophotometric determination of aliphatic aldehydes and for the determination of hexosamines in glycosaminoglycans. It is Incorporated into a new peroxidase color reaction. It is also a reagent for the spectrophotometric determination of traces of selenium (IV) in environmental samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a reagent for the spectrophotometric determination of aliphatic aldehydes and for the determination of hexosamines in glycosaminoglycans. It is Incorporated into a new peroxidase color reaction. It is also a reagent for the spectrophotometric determination of traces of selenium (IV) in environmental samples.
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents. Store at room temperature.
RUO – Research Use Only
General References:
  1. Mayumi Kaneko.; Makoto Kurihara.; Shigenori Nakano.; Takuji Kawashim. Flow-injection determination of chromium(III) by its catalysis on the oxidative coupling of 3-methyl-2-benzothiazolinone hydrazone with N-ethyl-N-(2-hydroxy-3-sulfopropyl)-3-methoxyaniline. Anal. Chim. Acta. 1998, 47 (5),1093-1098.
  2. Takashi Tomiyasu.; Norio Teshima.; Shigenori Nakano.; Takuji Kawashim. Kinetic-catalytic determination of traces of iron by the oxidative coupling reaction of 3-methyl-2-benzothiazolinone hydrazone with N,N-dimethylaniline. Talanta. 1998, 47 (5),1093-1098.