Sodium tetrakis(4-fluorophenyl)borate dihydrate, is used as a reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium tetrakis(4-fluorophenyl)borate dihydrate, is used as a reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.
Notes
Hygroscopic. Incompatible withstrong oxidizing agents.
RUO – Research Use Only
General References:
- Funada, T.; Hirose, T.; Tamai, N.; Yao, H. Organic nanoparticles of malachite green with enhanced far-red emission: size-dependence of particle rigidity. Phys. Chem. Chem. Phys. 2015, 17 (16), 11006-11013.
- Lisak, G.; Arnebrant, T.; Ruzgas, T.; Bobacka, J. Textile-based sampling for potentiometric determination of ions. Anal. Chim. Acta 2015, 877, 71-79.