2-Nitrofluorene, 98%, Thermo Scientific Chemicals
2-Nitrofluorene, 98%, Thermo Scientific Chemicals
2-Nitrofluorene, 98%, Thermo Scientific Chemicals
2-Nitrofluorene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Nitrofluorene, 98%, Thermo Scientific Chemicals
CAS: 607-57-8 | C13H9NO2 | 211.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFH55744.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Nitrofluorene
CAS607-57-8
Health Hazard 1H341-H351
Health Hazard 2GHS H Statement
H351
Suspected of causing cancer.
Health Hazard 3P201-P202-P281-P308+P313-P501c