Tri-n-butylphenyltin, 97%, Thermo Scientific Chemicals
Tri-n-butylphenyltin, 97%, Thermo Scientific Chemicals
Tri-n-butylphenyltin, 97%, Thermo Scientific Chemicals
Tri-n-butylphenyltin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tri-n-butylphenyltin, 97%, Thermo Scientific Chemicals
CAS: 960-16-7 | C18H32Sn | 367.164 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number ALFH55596.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTri-n-butylphenyltin
CAS960-16-7
Health Hazard 1H301-H312-H315-H319-H360FD-H372
Health Hazard 2GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P201-P202-P260-P264b-P270-P280i-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
View more
It is applied in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied in chemical research.
Solubility
Sparingly Soluble (9.9E-5 g/L) (25°C)
Notes
Store at room temperature. Avoid strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Dietmar. Seyferth.; Michael A. Weiner. The Preparation of Organolithium Compounds by the Transmetalation Reaction. IV. Some Factors Affecting the Transmetalation Reaction. J. Am. Chem. Soc. 1962, 84 (3),361-364.
  2. Dietmar Seyferth.; Michael Weiner. Communications: Preparation and Some Reactions of Allyllithium. J. Org. Chem. 1959, 24 (9),1395-1396.