3,5-Dibromoquinoline, 96%, Thermo Scientific Chemicals
3,5-Dibromoquinoline, 96%, Thermo Scientific Chemicals
3,5-Dibromoquinoline, 96%, Thermo Scientific Chemicals
3,5-Dibromoquinoline, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Dibromoquinoline, 96%, Thermo Scientific Chemicals
CAS: 101861-59-0 | C9H5Br2N | 286.954 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Catalog number ALFH54859.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material3,5-Dibromoquinoline
CAS101861-59-0
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c