trans-3-Heptene, 97%, Thermo Scientific Chemicals
trans-3-Heptene, 97%, Thermo Scientific Chemicals
trans-3-Heptene, 97%, Thermo Scientific Chemicals
trans-3-Heptene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-3-Heptene, 97%, Thermo Scientific Chemicals
CAS: 14686-14-7 | C7H14 | 98.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALFH53482.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS299-11-6
IUPAC Name5-methylphenazin-5-ium methyl sulfate
Molecular FormulaC14H14N2O4S
InChI KeyRXGJTUSBYWCRBK-UHFFFAOYSA-M
SMILESCOS([O-])(=O)=O.C[N+]1=C2C=CC=CC2=NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark yellow to khaki to brown
Appearance (Form)Crystalline powder
UV spectrophotometry>=97.5 % (0.01g/l H2O, 1cm cell, lambda max 387.5 nm)