trans-3-Heptene, 97%, Thermo Scientific Chemicals
trans-3-Heptene, 97%, Thermo Scientific Chemicals
trans-3-Heptene, 97%, Thermo Scientific Chemicals
trans-3-Heptene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-3-Heptene, 97%, Thermo Scientific Chemicals
CAS: 14686-14-7 | C7H14 | 98.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number ALFH53482.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7320-34-5
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO497.5 to 102.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder
Heavy metals (as Pb)=<10 ppm
Iron (Fe)=<10 ppm