trans-3-Heptene, 97%, Thermo Scientific Chemicals
trans-3-Heptene, 97%, Thermo Scientific Chemicals
trans-3-Heptene, 97%, Thermo Scientific Chemicals
trans-3-Heptene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-3-Heptene, 97%, Thermo Scientific Chemicals
CAS: 14686-14-7 | C7H14 | 98.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number ALFH53482.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Materialtrans-3-Heptene
CAS14686-14-7
Health Hazard 1H225-H304-H315-H319-H335
Health Hazard 2GHS H Statement
H225-H304-H315-H319-H335
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c