3-Ethynylpyridine, 97%, Thermo Scientific Chemicals
3-Ethynylpyridine, 97%, Thermo Scientific Chemicals
3-Ethynylpyridine, 97%, Thermo Scientific Chemicals
3-Ethynylpyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Ethynylpyridine, 97%, Thermo Scientific Chemicals
CAS: 2510-23-8 | C7H5N | 103.124 g/mol
Have Questions?
Catalog NumberQuantity
ALFH53465.031 g
Catalog number ALFH53465.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS95-73-8
IUPAC Name2,4-dichloro-1-methylbenzene
Molecular FormulaC7H6Cl2
InChI KeyFUNUTBJJKQIVSY-UHFFFAOYSA-N
SMILESCC1=CC=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5450 to 1.5470 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=98.5 %