N-Hydroxysulfosuccinimide sodium salt, 97%, Thermo Scientific Chemicals
N-Hydroxysulfosuccinimide sodium salt, 97%, Thermo Scientific Chemicals
N-Hydroxysulfosuccinimide sodium salt, 97%, Thermo Scientific Chemicals
N-Hydroxysulfosuccinimide sodium salt, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Hydroxysulfosuccinimide sodium salt, 97%, Thermo Scientific Chemicals
CAS: 106627-54-7 | C4H4NNaO6S | 217.127 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ALFH52795.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight217.13
Unit Size
N-Hydroxysulfosuccinimide sodium salt is used as crosslinking agents for preparing hydrophilic active esters. It is also used to activate carboxylic acids to the more reactive succinimidyl esters for coupling, biotinylation and conjugation with fluorophores. Further, it acts as a hydrophilic ligand and also used in covalently binding proteins to carboxyl containing beads, resins, other nanoparticles. It is involved in the conversion of carboxyl groups to primary amine-reactive sulfo-NHS esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Hydroxysulfosuccinimide sodium salt is used as crosslinking agents for preparing hydrophilic active esters. It is also used to activate carboxylic acids to the more reactive succinimidyl esters for coupling, biotinylation and conjugation with fluorophores. Further, it acts as a hydrophilic ligand and also used in covalently binding proteins to carboxyl containing beads, resins, other nanoparticles. It is involved in the conversion of carboxyl groups to primary amine-reactive sulfo-NHS esters.
Solubility
Soluble in methanol and water.
Notes
Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Neves, M. M. P. S.; González-García, M. B.; Hernández-Santos, D.; Fanjul-Bolado, P. Streptavidin functionalized nickel nanowires: A new ferromagnetic platform for biotinylated-based assays. Talanta 2015, 144, 283-288.
  2. Huang, X.; Xu, Z.; Mao, Y.; Ji, Y.; Xu, H.; Xiong, Y.; Li, Y. Gold nanoparticle-based dynamic light scattering immunoassay for ultrasensitive detection of Listeria monocytogenes in lettuces. Biosens. Bioelectron. 2015, 66, 184-190.s