Lovastatin, 97%, Thermo Scientific Chemicals
Lovastatin, 97%, Thermo Scientific Chemicals
Lovastatin, 97%, Thermo Scientific Chemicals
Lovastatin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lovastatin, 97%, Thermo Scientific Chemicals
CAS: 75330-75-5 | C24H36O5 | 404.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFH52792.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLovastatin
CAS75330-75-5
Health Hazard 1H317-H351-H361d
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more
Lovastatin is a cholesterol lowering drug and competitive inhibitor of HMG-CoA reductase, a rate limiting enzyme in cholesterol synthesis. Blocks the production of mevalonate, a critical compound in the production of cholesterol and isoprenoids. It increases cellular resistance to anticancer agents such as doxorubicin and induces apoptosis in myeloma cells. It causes cell cycle arrest in G1 and G2/M phases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Lovastatin is a cholesterol lowering drug and competitive inhibitor of HMG-CoA reductase, a rate limiting enzyme in cholesterol synthesis. Blocks the production of mevalonate, a critical compound in the production of cholesterol and isoprenoids. It increases cellular resistance to anticancer agents such as doxorubicin and induces apoptosis in myeloma cells. It causes cell cycle arrest in G1 and G2/M phases.
Solubility
Soluble in DMSO (10 mg/ml), ethanol (10 mg/ml), methanol, chloroform, acetone, acetonitrile, DMF, and water (0.4 µg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
RUO – Research Use Only
General References:
  1. Jonathan A.Tobert. Efficacy and long-term adverse effect pattern of lovastatin. The American Journal of Cardiology. 1988, 62, (15),J28-J34
  2. Peter Jones MD; Stephanie Kafonek MD; Irene Laurora PharmD; Donald Hunninghake MD. Comparative Dose Efficacy Study of Atorvastatin VersusSimvastatin, Pravastatin, Lovastatin, and Fluvastatin in Patients With Hypercholesterolemia (The CURVES Study). The American Journal of Cardiology. 1998, 81, (5),582-587