n-Butyllithium, 2.5M in hexane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals
n-Butyllithium, 2.5M in hexane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals
n-Butyllithium, 2.5M in hexane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals
n-Butyllithium, 2.5M in hexane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyllithium, 2.5M in hexane, packaged under Nitrogen in resealable AcroSeal™ bottles, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
Catalog number ALFH36949.AD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS223911-98-6
IUPAC Nameholmium(3+) tribromide
Molecular FormulaBr3Ho
InChI KeyMZNSYJWLQLXLHE-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[Ho+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.