2,4,6-Tris(3-pyridyl)-1,3,5-triazine, 97%, Thermo Scientific Chemicals
2,4,6-Tris(3-pyridyl)-1,3,5-triazine, 97%, Thermo Scientific Chemicals
2,4,6-Tris(3-pyridyl)-1,3,5-triazine, 97%, Thermo Scientific Chemicals
2,4,6-Tris(3-pyridyl)-1,3,5-triazine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4,6-Tris(3-pyridyl)-1,3,5-triazine, 97%, Thermo Scientific Chemicals
CAS: 42333-76-6 | C18H12N6 | 312.34 g/mol
Have Questions?
Catalog NumberQuantity
ALFH36052.031 g
Catalog number ALFH36052.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS580-13-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder or lumps
Assay (GC)≥98.0%