2-Aminooxazole, 97%, Thermo Scientific Chemicals
2-Aminooxazole, 97%, Thermo Scientific Chemicals
2-Aminooxazole, 97%, Thermo Scientific Chemicals
2-Aminooxazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminooxazole, 97%, Thermo Scientific Chemicals
CAS: 4570-45-0 | C3H4N2O | 84.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFH34360.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS114162-64-0
IUPAC Name(2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid; cyclohexanamine
Molecular FormulaC20H26BrClN2O7
InChI KeyJXCKZXHCJOVIAV-VLQIOUPANA-N
SMILESNC1CCCCC1.O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)O[C@@H]([C@H]1O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or needles
HPLC>=97.5 %
Appearance (Color)Colorless to white to off-white
Infrared spectrumConforms
Water=<5 % (K.F.)