5-(4-Chlorophenyl)-1,3,4-oxadiazole-2-thiol, 96%
5-(4-Chlorophenyl)-1,3,4-oxadiazole-2-thiol, 96%
5-(4-Chlorophenyl)-1,3,4-oxadiazole-2-thiol, 96%
5-(4-Chlorophenyl)-1,3,4-oxadiazole-2-thiol, 96%
5-(4-Chlorophenyl)-1,3,4-oxadiazole-2-thiol, 96%
CAS: 23766-28-1 | C8H5ClN2OS | 212.65 g/mol
Have Questions?
Catalog NumberQuantity
ALFH33429.065 g
Catalog number ALFH33429.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material5-(4-Chlorophenyl)-1,3,4-oxadiazole-2-thiol
Melting Point169°C to 170°C
CAS23766-28-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement