Filter aid, Celite∣r 545, treated with sodium carbonate, flux calcined, Thermo Scientific Chemicals
Filter aid, Celite∣r 545, treated with sodium carbonate, flux calcined, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Filter aid, Celite∣r 545, treated with sodium carbonate, flux calcined, Thermo Scientific Chemicals
CAS: 68855-54-9 | CNa2O3 | 105.988 g/mol
Have Questions?
Catalog NumberQuantity
ALFH33152.0B1000 g
Catalog number ALFH33152.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS7647-15-6
IUPAC Namesodium bromide
Molecular FormulaBrNa
InChI KeyJHJLBTNAGRQEKS-UHFFFAOYSA-M
SMILES[Na+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=99.5 %
Nitrogen compounds (as N)=<0.005 %
Iodide (I)=<50 ppm
Magnesium (Mg)=<50 ppm
Nickel (Ni)=<10 ppm
View more
Celite filter aids maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization. A medium to fine product for the filtration of most process streams including beer, cider, wines, water, low viscosity glues and organic chemicals and pharmaceuticals. for water purification.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Celite filter aids maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization. A medium to fine product for the filtration of most process streams including beer, cider, wines, water, low viscosity glues and organic chemicals and pharmaceuticals. for water purification.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong acids.
RUO – Research Use Only
General References:
  1. J A Zeevaart. Effects of photoperiod on growth rate and endogenous gibberellins in the long-day rosette plant spinach. Plant Physiology. 197, 1971(6), 821-827.
  2. N CITRI et. al.The effect of urea and guanidine hydrochloride on activity and optical rotation of penicillinase.. Journal of Biological Chemistry. 1960, 235 3454-3459.