1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals
1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals
1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals
1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals
CAS: 84322-56-5 | C6F3I3 | 509.775 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFH33116.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS554-01-8
IUPAC Name6-amino-5-methyl-1,2-dihydropyrimidin-2-one
Molecular FormulaC5H7N3O
InChI KeyLRSASMSXMSNRBT-UHFFFAOYSA-N
SMILESCC1=C(N)NC(=O)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)> 96.0%
Appearance (Color)White to cream
FormPowder
1,3,5-Trifluoro-2,4,6-triiodobenzene is useful for biological applications. Further, it is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3,5-Trifluoro-2,4,6-triiodobenzene is useful for biological applications. Further, it is used as an intermediate in organic synthesis.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only
General References:
  1. Lieffrig, J.; Jeannin, O.; Fourmigué, M. Expanded Halogen-Bonded Anion Organic Networks with Star-Shaped Iodoethynyl-Substituted Molecules: From Corrugated 2D Hexagonal Lattices to Pyrite-Type 2-Fold Interpenetrated Cubic Lattices. J. Am. Chem. Soc. 2013, 135 (16), 6200-6210.
  2. Ji, B.; Zhang, Y.; Deng, D.; Wang, W. Improper halogen bond in the crystal structure. CrystEngComm 2013, 15 (16), 3093-3096.