Amberlite™ IRA-96, ion exchange resin, Thermo Scientific Chemicals
Amberlite™ IRA-96, ion exchange resin, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Amberlite™ IRA-96, ion exchange resin, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
Catalog number ALFH32475.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS65719-09-7
IUPAC Namemethyl 2-methylpyridine-3-carboxylate
Molecular FormulaC8H9NO2
InChI KeyKLHWBYHFWALOIJ-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(C)N=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Infrared spectrumConforms
Water=<1.5 %
Refractive index1.5145 to 1.5185 (20°C, 589 nm)