Celatom∣r FW-14, Thermo Scientific Chemicals
Celatom∣r FW-14, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Celatom∣r FW-14, Thermo Scientific Chemicals
CAS: 68855-54-9 | CNa2O3 | 105.988 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
Catalog number ALFH31733.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCelatom™ FW-14
Quantity1000 g
CAS68855-54-9
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H351-H373
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.