Furan-2,5-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Furan-2,5-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Furan-2,5-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Furan-2,5-dicarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Furan-2,5-dicarboxylic acid, 98%, Thermo Scientific Chemicals
CAS: 3238-40-2 | C6H4O5 | 156.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFH28718.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS68758-68-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to cream
FormPowder or crystalline powder
Melting Point (clear melt)126-135?C