Ethyl 7-bromoheptanoate, 97%
Ethyl 7-bromoheptanoate, 97%
Ethyl 7-bromoheptanoate, 97%
Ethyl 7-bromoheptanoate, 97%
Thermo Scientific Chemicals
Ethyl 7-bromoheptanoate, 97%
CAS: 29823-18-5 | C9H17BrO2 | 237.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFH28001.AA5 mL
Catalog number ALFH28001.AA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl 7-bromoheptanoate
CAS29823-18-5
Health Hazard 1H315-H317-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c