2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde, 96%, Thermo Scientific Chemicals
2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde, 96%, Thermo Scientific Chemicals
2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde, 96%, Thermo Scientific Chemicals
2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde, 96%, Thermo Scientific Chemicals
2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde, 96%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFH27844.031 g
Catalog number ALFH27844.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde
CAS5604-46-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c