Ethyl cellulose
Ethyl cellulose
Ethyl cellulose
Thermo Scientific Chemicals
Ethyl cellulose
CAS: 9004-57-3 | C23H24N6O4 | 448.483 g/mol
Have Questions?
Catalog NumberQuantity
ALFH27621.MD250 mg
Catalog number ALFH27621.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS594-70-7
IUPAC Name2-methyl-2-nitropropane
Molecular FormulaC4H9NO2
InChI KeyAIMREYQYBFBEGQ-UHFFFAOYSA-N
SMILESCC(C)(C)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
FormFused solid or clear liquid as melt
Assay (GC)≥98.0%
Refractive Index1.3970-1.4015 @ 30?C
Ethyl cellulose is used as a food additive and as an emulsifier. It is also used as a film-former in coatings, hot-melt adhesives and transfer inks. Further, it is employed as a binder, filler, plastic coating and diluents of color. In addition to this, it is used in lacquers and plastic toughing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl cellulose is used as a food additive and as an emulsifier. It is also used as a film-former in coatings, hot-melt adhesives and transfer inks. Further, it is employed as a binder, filler, plastic coating and diluents of color.
RUO – Research Use Only
General References:
  1. Davidovich-Pinhas, M.; Barbut, S.; Marangon, A. G. The gelation of oil using ethyl cellulose. Carbohydr. Polym. 2015, 117, 869-878.
  2. Djerafi, R.; Masmoudi, Y.; Crampon, C.; Meniai, A.; Badens, E. Supercritical anti-solvent precipitation of ethyl cellulose. J. Supercrit. Fluids 2015, 105, 92-98.