4-Iodobiphenyl, 97%, Thermo Scientific Chemicals
4-Iodobiphenyl, 97%, Thermo Scientific Chemicals
4-Iodobiphenyl, 97%, Thermo Scientific Chemicals
4-Iodobiphenyl, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Iodobiphenyl, 97%, Thermo Scientific Chemicals
CAS: 1591-31-7 | C12H9I | 280.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFH27453.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1941-26-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellowish-white
Appearance (Form)Crystalline powder
Titration with HClO497.5 to 102.5 %
Infrared spectrumConforms
4-Iodobiphenyl is used as a starting material for the preparation of p-terphenyl derivatives. It is also used in Suzuki reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Iodobiphenyl is used as a starting material for the preparation of p-terphenyl derivatives. It is also used in Suzuki reaction.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only
General References:
  1. Okoye, N. H.; de Silva, U. K.; Wengatz, J. A.; Lapitsky, Y. Photodirected assembly of polyelectrolyte complexes. Polymer 2015, 60, 69-76.
  2. Liu, W.; Xu, L. Ligand-controlled chemoselectivity in potassium tert-amylate-promoted direct C-H arylation of unactivated benzene with aryl halides. Tetrahedron 2015, 71 (30), 4974-4981.