1-Methyl-3-phenylpiperazine, 97%, Thermo Scientific Chemicals
1-Methyl-3-phenylpiperazine, 97%, Thermo Scientific Chemicals
1-Methyl-3-phenylpiperazine, 97%, Thermo Scientific Chemicals
1-Methyl-3-phenylpiperazine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-3-phenylpiperazine, 97%, Thermo Scientific Chemicals
CAS: 5271-27-2 | C11H16N2 | 176.263 g/mol
Have Questions?
Catalog NumberQuantity
ALFH26924.031 g
Catalog number ALFH26924.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS21640-48-2
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
FormPowder
Melting Point (clear melt)66.5-75.5?C
Appearance (Color)White
Assay (HPLC)≥95.0%