2',4,4'-Trihydroxychalcone, 97%, Thermo Scientific™
2',4,4'-Trihydroxychalcone, 97%, Thermo Scientific™
2',4,4'-Trihydroxychalcone, 97%, Thermo Scientific™
2',4,4'-Trihydroxychalcone, 97%, Thermo Scientific™
2',4,4'-Trihydroxychalcone, 97%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFH26920.031 g
Catalog number ALFH26920.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material2',4,4'-Trihydroxychalcone
CAS961-29-5
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.