Hydrogen chloride, 2M in diethyl ether, Thermo Scientific Chemicals
Hydrogen chloride, 2M in diethyl ether, Thermo Scientific Chemicals
Hydrogen chloride, 2M in diethyl ether, Thermo Scientific Chemicals
Hydrogen chloride, 2M in diethyl ether, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrogen chloride, 2M in diethyl ether, Thermo Scientific Chemicals
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantity
ALFH26914.AP500 mL
Catalog number ALFH26914.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS1978-33-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
Assay from Suppliers CofA≥96.0%
FormLiquid
Hydrogen chloride, 2M in diethyl ether is used in organic synthesis to prepare emodin derivatives and polyamine derivatives of betulinic acid and beta-sitosterol.