4'-n-Octylbiphenyl-4-carbonitrile, 99%, Thermo Scientific Chemicals
4'-n-Octylbiphenyl-4-carbonitrile, 99%, Thermo Scientific Chemicals
4'-n-Octylbiphenyl-4-carbonitrile, 99%, Thermo Scientific Chemicals
4'-n-Octylbiphenyl-4-carbonitrile, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4'-n-Octylbiphenyl-4-carbonitrile, 99%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFH26318.031 g
Catalog number ALFH26318.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material4'-n-Octylbiphenyl-4-carbonitrile
CAS52709-84-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
The thermobarometry of thermotropic liquid crystals is feasible by the metal tube Berthelot method, and its extension in the negative-pressure range will be interesting for the study of organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The thermobarometry of thermotropic liquid crystals is feasible by the metal tube Berthelot method, and its extension in the negative-pressure range will be interesting for the study of organic compounds.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
RUO – Research Use Only
General References:
  1. Yoshihito Ohde,; Yasutoshi Tanzawa,; Kaname Motoshita,; Kazuki Hiros.Thermobarometry for 4'-n-Octylbiphenyl-4-carbonitrile in Metal Tube Berthelot Method and Polymorphism in Crystalline Phase of 4'-n-Octylbiphenyl-4-carbonitrile Found through Cooling Paths in Negative-Pressure Range. Japanese Journal of Applied Physics. 2008 , 47 7R