5-Chloro-2-fluorobenzaldehyde, 97%, Thermo Scientific Chemicals
5-Chloro-2-fluorobenzaldehyde, 97%, Thermo Scientific Chemicals
5-Chloro-2-fluorobenzaldehyde, 97%, Thermo Scientific Chemicals
5-Chloro-2-fluorobenzaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Chloro-2-fluorobenzaldehyde, 97%, Thermo Scientific Chemicals
CAS: 96515-79-6 | C7H4ClFO | 158.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFH26187.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS50-00-0
IUPAC Nameformaldehyde
Molecular FormulaCH2O
InChI KeyWSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILESC=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH7.40 ± 0.20
Appearance (Color)Clear colorless
FormLiquid
CommentFormaldehyde solution is made by dissolving it in 1X PBS, pH 7.4, followed by submicron filtration