2-Fluoro-6-methoxybenzoic acid, 97%, Thermo Scientific™
2-Fluoro-6-methoxybenzoic acid, 97%, Thermo Scientific™
2-Fluoro-6-methoxybenzoic acid, 97%, Thermo Scientific™
2-Fluoro-6-methoxybenzoic acid, 97%, Thermo Scientific™
2-Fluoro-6-methoxybenzoic acid, 97%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFH26060.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Fluoro-6-methoxybenzoic acid
CAS137654-21-8
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.