Albendazole, 98+%, Thermo Scientific Chemicals
Albendazole, 98+%, Thermo Scientific Chemicals
Albendazole, 98+%, Thermo Scientific Chemicals
Albendazole, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Albendazole, 98+%, Thermo Scientific Chemicals
CAS: 54965-21-8 | C12H15N3O2S | 265.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
Catalog number ALFH25925.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS329-98-6
IUPAC Namephenylmethanesulfonyl fluoride
Molecular FormulaC7H7FO2S
InChI KeyYBYRMVIVWMBXKQ-UHFFFAOYSA-N
SMILESFS(=O)(=O)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Needles and/or agglomerates
Melting point90°C to 94°C
Appearance (Color)White
Infrared spectrumConforms
GC>=98.5 %