Albendazole, 98+%, Thermo Scientific Chemicals
Albendazole, 98+%, Thermo Scientific Chemicals
Albendazole, 98+%, Thermo Scientific Chemicals
Albendazole, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Albendazole, 98+%, Thermo Scientific Chemicals
CAS: 54965-21-8 | C12H15N3O2S | 265.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Catalog number ALFH25925.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS530-48-3
IUPAC Name(1-phenylethenyl)benzene
Molecular FormulaC14H12
InChI KeyZMYIIHDQURVDRB-UHFFFAOYSA-N
SMILESC=C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.6045-1.6095 @ 20?C
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%