5-Bromo-2-hydroxybenzophenone, 97%, Thermo Scientific Chemicals
5-Bromo-2-hydroxybenzophenone, 97%, Thermo Scientific Chemicals
5-Bromo-2-hydroxybenzophenone, 97%, Thermo Scientific Chemicals
5-Bromo-2-hydroxybenzophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Bromo-2-hydroxybenzophenone, 97%, Thermo Scientific Chemicals
CAS: 55082-33-2 | C13H9BrO2 | 277.117 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFB25507.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS108-24-7
IUPAC Nameacetyl acetate
Molecular FormulaC4H6O3
InChI KeyWFDIJRYMOXRFFG-UHFFFAOYSA-N
SMILESCC(=O)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Chloride (Cl)=<5 ppm
Sulfate (SO4)=<5 ppm
Calcium (Ca)=<0.5 ppm
Copper (Cu)=<0.02 ppm